Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 15.48 mm, Tool Steel
Catalog No. 5000527127
Encompass
This pin gage is designed for precise measurement of hole sizes. It is constructed from durable tool steel with a hardness of Rc 60/65 and features a steel finish of 10 microfinish or better. With a diameter of 15.48 mm and an overall length of 2 in, this gage provides accurate measurements within a tolerance of +.000/-.005 in.
  • Measures hole sizes within +.000/-.005 in.
  • Includes certificate of accuracy with NIST traceability.
Catalog No. 50-005-27127 Supplier MSC Supplier No. 40442931
This pin gage is designed for precise measurement of hole sizes. It is constructed from durable tool steel with a hardness of Rc 60/65 and features a steel finish of 10 microfinish or better. With a diameter of 15.48 mm and an overall length of 2 in, this gage provides accurate measurements within a tolerance of +.000/-.005 in.
  • Measures hole sizes within +.000/-.005 in.
  • Includes certificate of accuracy with NIST traceability.