Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 19.32 mm
Catalog No. 5000527132
Encompass
This pin gage is designed for precise measurement of hole sizes. It is a Class ZZ Minus pin gage, made from durable tool steel, providing reliable accuracy for quality control and inspection tasks.
  • Pin gage type
  • Minus tolerance class ZZ
  • Manufactured from tool steel with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
  • 19.32 mm diameter
  • Tolerance: +.000/-.005 in
  • 2 in overall length
  • Measures hole sizes within +.000/-.005 in
  • Includes certificate of accuracy with NIST traceability
  • Package quantity of 1
Catalog No. 50-005-27132 Supplier MSC Supplier No. 40446775
This pin gage is designed for precise measurement of hole sizes. It is a Class ZZ Minus pin gage, made from durable tool steel, providing reliable accuracy for quality control and inspection tasks.
  • Pin gage type
  • Minus tolerance class ZZ
  • Manufactured from tool steel with Rc 60/65 hardness
  • Steel finish with 10 microfinish or better
  • 19.32 mm diameter
  • Tolerance: +.000/-.005 in
  • 2 in overall length
  • Measures hole sizes within +.000/-.005 in
  • Includes certificate of accuracy with NIST traceability
  • Package quantity of 1