Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 2.83 mm, Tool Steel
Catalog No. 5000527022
Encompass
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with an Rc 60/65 hardness, it features a 2.83 mm diameter, a Class ZZ tolerance of +.000/-.005, and an overall length of 2 inches. The gage has a steel finish with 10 microfinish or better, ensuring accuracy and reliability.
  • Measures hole sizes within +.000/-.005".
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel with Rc 60/65 hardness.
  • Features a steel finish with 10 microfinish or better.
  • Class ZZ tolerance for high precision.
  • NoGo (minus) type for specific measurements.
Catalog No. 50-005-27022 Supplier MSC Supplier No. 40430282
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with an Rc 60/65 hardness, it features a 2.83 mm diameter, a Class ZZ tolerance of +.000/-.005, and an overall length of 2 inches. The gage has a steel finish with 10 microfinish or better, ensuring accuracy and reliability.
  • Measures hole sizes within +.000/-.005".
  • Includes certificate of accuracy with NIST traceability.
  • Made from tool steel with Rc 60/65 hardness.
  • Features a steel finish with 10 microfinish or better.
  • Class ZZ tolerance for high precision.
  • NoGo (minus) type for specific measurements.