Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 20.46 mm, Tool Steel
Catalog No. 5000533593
Encompass
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with an Rc 60/65 hardness, it features a 10 microfinish or better for accuracy. With a diameter of 20.46 mm and a tolerance of +.000/-.005, this minus (NoGo) pin gage has an overall length of 2 in.
  • Constructed from tool steel with Rc 60/65 hardness
  • Features a tolerance of +.000/-.005
  • Measures hole sizes within +.000/-.005"
  • Includes a certificate of accuracy with NIST traceability
  • Has an overall length of 2 in
  • Designed as a minus (NoGo) type
  • Conforms to class ZZ standards
  • Finished with a 10 microfinish or better
Catalog No. 50-005-33593 Supplier MSC Supplier No. 40447914
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel with an Rc 60/65 hardness, it features a 10 microfinish or better for accuracy. With a diameter of 20.46 mm and a tolerance of +.000/-.005, this minus (NoGo) pin gage has an overall length of 2 in.
  • Constructed from tool steel with Rc 60/65 hardness
  • Features a tolerance of +.000/-.005
  • Measures hole sizes within +.000/-.005"
  • Includes a certificate of accuracy with NIST traceability
  • Has an overall length of 2 in
  • Designed as a minus (NoGo) type
  • Conforms to class ZZ standards
  • Finished with a 10 microfinish or better