Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 6.51 mm, Tool Steel
Catalog No. 5000536164
Encompass
The Vermont Gage pin gage is a precision measuring tool crafted from durable tool steel with a hardness of Rc 60/65. Designed to accurately measure hole sizes, this Class ZZ Minus type gage has a 6.51 mm diameter and an overall length of 2 inches. It is ideal for applications requiring precise dimensional verification and comes with a Certificate of Accuracy with NIST Traceability.
  • Made from tool steel with Rc 60/65 hardness
  • Steel finish of 10 microfinish or better
  • Diameter of 6.51 mm
  • Overall length of 2 in
  • NoGo (minus) type
  • Class ZZ gage
  • Tolerance of +.000/-.005
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-36164 Supplier MSC Supplier No. 40433963
The Vermont Gage pin gage is a precision measuring tool crafted from durable tool steel with a hardness of Rc 60/65. Designed to accurately measure hole sizes, this Class ZZ Minus type gage has a 6.51 mm diameter and an overall length of 2 inches. It is ideal for applications requiring precise dimensional verification and comes with a Certificate of Accuracy with NIST Traceability.
  • Made from tool steel with Rc 60/65 hardness
  • Steel finish of 10 microfinish or better
  • Diameter of 6.51 mm
  • Overall length of 2 in
  • NoGo (minus) type
  • Class ZZ gage
  • Tolerance of +.000/-.005
  • Includes certificate of accuracy with NIST traceability