Learn More
MSC Vermont Gage Pin Gage, Minus, Class ZZ, 7.89 mm, Tool Steel
Catalog No. 5000527158
Encompass
This pin gage is a precision measuring tool designed to measure hole sizes. It is constructed from durable tool steel with a hardness rating of Rc 60/65 and features a steel finish with 10 microinch or better for accuracy and longevity. This NoGo (Minus) gage has a diameter of 7.89 mm and an overall length of 2 inches. It comes with a Certificate of Accuracy with NIST traceability, ensuring its calibration and precision.
  • Made from tool steel, Rc 60/65 hardness.
  • Measures hole sizes within a tolerance of +.000/-.005 "
  • Includes a Certificate of Accuracy with NIST traceability.
  • Overall length of 2 inches.
  • Minus type (NoGo) pin gage.
  • Steel finish with 10 microinch or better.
  • Diameter of 7.89 mm.
  • Class ZZ precision.
  • Made in the United States.
Catalog No. 50-005-27158 Supplier MSC Supplier No. 40435349
This pin gage is a precision measuring tool designed to measure hole sizes. It is constructed from durable tool steel with a hardness rating of Rc 60/65 and features a steel finish with 10 microinch or better for accuracy and longevity. This NoGo (Minus) gage has a diameter of 7.89 mm and an overall length of 2 inches. It comes with a Certificate of Accuracy with NIST traceability, ensuring its calibration and precision.
  • Made from tool steel, Rc 60/65 hardness.
  • Measures hole sizes within a tolerance of +.000/-.005 "
  • Includes a Certificate of Accuracy with NIST traceability.
  • Overall length of 2 inches.
  • Minus type (NoGo) pin gage.
  • Steel finish with 10 microinch or better.
  • Diameter of 7.89 mm.
  • Class ZZ precision.
  • Made in the United States.