Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 0.2625 in, Tool Steel
Catalog No. 5000510462
Encompass
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a hardness of Rc 60/65 and a 10 microfinish or better, it ensures accurate readings within a tolerance of +.0002/-.0000. Each gage has an overall length of 2 inches and comes with a certificate of accuracy traceable to NIST.
  • Made from tool steel with Rc 60/65 hardness
  • Maintains a tolerance of +.0002/-.0000
  • Measures hole sizes with high precision
  • Includes a certificate of accuracy with NIST traceability
  • Features an overall length of 2 inches
  • Has a 0.2625 inch diameter
  • Finished to 10 microfinish or better
  • Designed as a Go (Plus) type gage
  • Meets Class ZZ specifications
Catalog No. 50-005-10462 Supplier MSC Supplier No. 88926258
This pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a hardness of Rc 60/65 and a 10 microfinish or better, it ensures accurate readings within a tolerance of +.0002/-.0000. Each gage has an overall length of 2 inches and comes with a certificate of accuracy traceable to NIST.
  • Made from tool steel with Rc 60/65 hardness
  • Maintains a tolerance of +.0002/-.0000
  • Measures hole sizes with high precision
  • Includes a certificate of accuracy with NIST traceability
  • Features an overall length of 2 inches
  • Has a 0.2625 inch diameter
  • Finished to 10 microfinish or better
  • Designed as a Go (Plus) type gage
  • Meets Class ZZ specifications