Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 0.747 Inch
Catalog No. 5000531701
Encompass
This pin gage is a high-precision tool for accurately determining hole sizes. Crafted from robust tool steel (Rc 60/65) with a fine finish, it ensures reliable measurements. As a "Go (Plus)" type gage, it checks if a hole meets the minimum diameter. Each gage includes a Certificate of Accuracy with NIST Traceability, guaranteeing adherence to national standards.
  • High-quality tool steel (Rc 60/65) for durability.
  • Precision 10 microfinish or better for accurate measurements.
  • Tight tolerance of +.0002/-.0000 for accurate hole verification.
  • NIST traceable certificate of accuracy.
  • Optimal 2 inch length for handling.
  • "Go (Plus)" type verifies minimum hole size.
  • Class ZZ accuracy for manufacturing precision.
Catalog No. 50-005-31701 Supplier MSC Supplier No. 88974704
This pin gage is a high-precision tool for accurately determining hole sizes. Crafted from robust tool steel (Rc 60/65) with a fine finish, it ensures reliable measurements. As a "Go (Plus)" type gage, it checks if a hole meets the minimum diameter. Each gage includes a Certificate of Accuracy with NIST Traceability, guaranteeing adherence to national standards.
  • High-quality tool steel (Rc 60/65) for durability.
  • Precision 10 microfinish or better for accurate measurements.
  • Tight tolerance of +.0002/-.0000 for accurate hole verification.
  • NIST traceable certificate of accuracy.
  • Optimal 2 inch length for handling.
  • "Go (Plus)" type verifies minimum hole size.
  • Class ZZ accuracy for manufacturing precision.