Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 0.978 Inch
Catalog No. 5000530231
Encompass
This Vermont Gage pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a Rc 60/65 hardness, it features a steel finish with 10 microfinish or better. This Class ZZ pin gage has a diameter of 0.9780 inch and a plus tolerance of +.0002/-.0000. It comes with a Certificate of Accuracy with NIST Traceability.
  • Made from tool steel with Rc 60/65 hardness
  • Has a tolerance of +.0002/-.0000
  • Measures hole sizes within +.0002/-.0000
  • Includes a Certificate of Accuracy with NIST traceability
  • Features an overall length of 2 in
  • Is a Go (Plus) type
  • Has a steel finish with 10 microfinish or better
  • Belongs to Class ZZ
  • Manufactured in the United States
Catalog No. 50-005-30231 Supplier MSC Supplier No. 88997804
This Vermont Gage pin gage is designed for precise measurement of hole sizes. Made from durable tool steel with a Rc 60/65 hardness, it features a steel finish with 10 microfinish or better. This Class ZZ pin gage has a diameter of 0.9780 inch and a plus tolerance of +.0002/-.0000. It comes with a Certificate of Accuracy with NIST Traceability.
  • Made from tool steel with Rc 60/65 hardness
  • Has a tolerance of +.0002/-.0000
  • Measures hole sizes within +.0002/-.0000
  • Includes a Certificate of Accuracy with NIST traceability
  • Features an overall length of 2 in
  • Is a Go (Plus) type
  • Has a steel finish with 10 microfinish or better
  • Belongs to Class ZZ
  • Manufactured in the United States