Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 16.53 mm, Tool Steel
Catalog No. 5000536312
Encompass
This pin gage is designed for precise measurement of hole sizes. It is a Plus, Class ZZ type, featuring a 16.53 mm diameter and constructed from durable tool steel.
  • Measures hole sizes with a tolerance of +.005/-.000.
  • Includes a certificate of accuracy with NIST traceability.
  • Features an overall length of 2 in.
  • Manufactured with a steel finish of 10 microfinish or better.
  • Made from tool steel with Rc 60/65 hardness.
  • Designed for 'Go' inspection applications.
Catalog No. 50-005-36312 Supplier MSC Supplier No. 68107580
This pin gage is designed for precise measurement of hole sizes. It is a Plus, Class ZZ type, featuring a 16.53 mm diameter and constructed from durable tool steel.
  • Measures hole sizes with a tolerance of +.005/-.000.
  • Includes a certificate of accuracy with NIST traceability.
  • Features an overall length of 2 in.
  • Manufactured with a steel finish of 10 microfinish or better.
  • Made from tool steel with Rc 60/65 hardness.
  • Designed for 'Go' inspection applications.