Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 16.56 mm, Tool Steel
Catalog No. 5000531855
Encompass
This Vermont Gage pin gage is a precision measuring tool designed for accurately determining hole sizes. Crafted from durable tool steel with a high-quality finish, it offers reliable performance for a range of applications.
  • Measures hole sizes within +.005/-.000 inch tolerance.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a finish of 10 microfinish or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed as a Class ZZ "Go" (Plus) type gage.
  • Has an overall length of 2 inches.
Catalog No. 50-005-31855 Supplier MSC Supplier No. 68107614
This Vermont Gage pin gage is a precision measuring tool designed for accurately determining hole sizes. Crafted from durable tool steel with a high-quality finish, it offers reliable performance for a range of applications.
  • Measures hole sizes within +.005/-.000 inch tolerance.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Features a finish of 10 microfinish or better.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed as a Class ZZ "Go" (Plus) type gage.
  • Has an overall length of 2 inches.