Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 16.95 mm, Tool Steel
Catalog No. 5000537310
Encompass
This pin gage is constructed from tool steel and designed for precise measurement of hole sizes. It features a Class ZZ, Go (Plus) type, with a tight tolerance of +.005/-.000. The gage has a steel finish with a 10 microfinish or better and an overall length of 2 in. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Designed for measuring hole sizes within a tolerance of +.005/-.000.
  • Features a steel finish with 10 microfinish or better.
  • Overall length of 2 in.
  • Go (Plus) type, Class ZZ.
  • Includes a certificate of accuracy with NIST traceability.
  • Made in the United States.
Catalog No. 50-005-37310 Supplier MSC Supplier No. 68108000
This pin gage is constructed from tool steel and designed for precise measurement of hole sizes. It features a Class ZZ, Go (Plus) type, with a tight tolerance of +.005/-.000. The gage has a steel finish with a 10 microfinish or better and an overall length of 2 in. Each gage includes a Certificate of Accuracy with NIST traceability.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Designed for measuring hole sizes within a tolerance of +.005/-.000.
  • Features a steel finish with 10 microfinish or better.
  • Overall length of 2 in.
  • Go (Plus) type, Class ZZ.
  • Includes a certificate of accuracy with NIST traceability.
  • Made in the United States.