Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 18.39mm
Catalog No. 5000527343
Encompass
This precision pin gage is ideal for accurately measuring hole sizes. Constructed from durable tool steel with a hardness of Rc 60/65, it features a high-quality steel finish.
  • Made from tool steel with Rc 60/65 hardness
  • Features a steel finish with 10 microfinish or better
  • Designed with a diameter of 18.39 mm
  • Provides a tolerance of +.005/-.000 in
  • Overall length of 2 in (50.8 mm)
  • Go (Plus) type
  • Class ZZ for precision applications
  • Includes a certificate of accuracy with NIST traceability
  • Measures hole sizes within +.005/-.000 in
Catalog No. 50-005-27343 Supplier MSC Supplier No. 68109446
This precision pin gage is ideal for accurately measuring hole sizes. Constructed from durable tool steel with a hardness of Rc 60/65, it features a high-quality steel finish.
  • Made from tool steel with Rc 60/65 hardness
  • Features a steel finish with 10 microfinish or better
  • Designed with a diameter of 18.39 mm
  • Provides a tolerance of +.005/-.000 in
  • Overall length of 2 in (50.8 mm)
  • Go (Plus) type
  • Class ZZ for precision applications
  • Includes a certificate of accuracy with NIST traceability
  • Measures hole sizes within +.005/-.000 in