Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 19.89 mm
Catalog No. 5000527395
Encompass
This pin gage is made from tool steel with a hardness of Rc 60/65 and a steel finish of 10 microinch or better. It has a diameter of 19.89 mm and an overall length of 2 in (50.8 mm). This 'Go' (Plus) class ZZ gage is designed to measure hole sizes within a tolerance of +.005/-.000 in.
  • Made from tool steel with a hardness of Rc 60/65.
  • Features a steel finish of 10 microinch or better.
  • Tolerance of +.005/-.000 in.
  • Measures hole sizes within +.005/-.000 in.
  • Overall length of 2 in (50.8 mm).
  • Includes a certificate of accuracy with NIST traceability.
  • Country of origin: United States.
Catalog No. 50-005-27395 Supplier MSC Supplier No. 68110949
This pin gage is made from tool steel with a hardness of Rc 60/65 and a steel finish of 10 microinch or better. It has a diameter of 19.89 mm and an overall length of 2 in (50.8 mm). This 'Go' (Plus) class ZZ gage is designed to measure hole sizes within a tolerance of +.005/-.000 in.
  • Made from tool steel with a hardness of Rc 60/65.
  • Features a steel finish of 10 microinch or better.
  • Tolerance of +.005/-.000 in.
  • Measures hole sizes within +.005/-.000 in.
  • Overall length of 2 in (50.8 mm).
  • Includes a certificate of accuracy with NIST traceability.
  • Country of origin: United States.