Learn More
MSC Vermont Gage Pin Gage, Plus, Class ZZ, 6.83mm, Tool Steel
Catalog No. 5000510657
Encompass
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel, it offers reliable performance and includes a Certificate of Accuracy with NIST traceability for verified precision.
  • Measures hole sizes within specified tolerance
  • Includes certificate of accuracy with NIST traceability
  • Go (Plus) type
  • Class ZZ
  • Steel finish with 10 microfinish or better
  • Tool steel material with Rc 60/65 hardness
  • 2 in overall length
  • 6.83 mm diameter
Catalog No. 50-005-10657 Supplier MSC Supplier No. 68097880
This pin gage is designed for precise measurement of hole sizes. Constructed from durable tool steel, it offers reliable performance and includes a Certificate of Accuracy with NIST traceability for verified precision.
  • Measures hole sizes within specified tolerance
  • Includes certificate of accuracy with NIST traceability
  • Go (Plus) type
  • Class ZZ
  • Steel finish with 10 microfinish or better
  • Tool steel material with Rc 60/65 hardness
  • 2 in overall length
  • 6.83 mm diameter