Learn More
MSC Vermont Gage Pin Gage Set, Class ZZ, Metric, Plus Tolerance, Steel
Catalog No. 5000511590
Encompass
This economical general purpose pin gage set is designed for quick and accurate measurements of hole sizes, distances between holes, slot widths, and Go/NoGo gaging. It features a Class ZZ tolerance for precise results and is suitable for various laboratory and industrial applications.
  • Class ZZ tolerance (.005 mm)
  • Go (plus) tolerance
  • Constructed from tool steel (60/65 Rc)
  • 2 in long
  • Surface finish of 10 microinch or better
  • Roundness within .0025 mm
  • Gages marked with size and tolerance direction
  • NIST traceable with included Certificate of Accuracy
  • Set includes 55 gages with .02 mm increments
Catalog No. 50-005-11590 Supplier MSC Supplier No. 40436248
This economical general purpose pin gage set is designed for quick and accurate measurements of hole sizes, distances between holes, slot widths, and Go/NoGo gaging. It features a Class ZZ tolerance for precise results and is suitable for various laboratory and industrial applications.
  • Class ZZ tolerance (.005 mm)
  • Go (plus) tolerance
  • Constructed from tool steel (60/65 Rc)
  • 2 in long
  • Surface finish of 10 microinch or better
  • Roundness within .0025 mm
  • Gages marked with size and tolerance direction
  • NIST traceable with included Certificate of Accuracy
  • Set includes 55 gages with .02 mm increments