Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 15.17 mm, Class ZZ, Minus
Catalog No. 5000532845
Encompass
Vermont Gage Pin Gages are economical, general-purpose tools for quick and accurate measurements of hole sizes, distances, and slot widths. They are also suitable for Go/NoGo gaging. This 15.17 mm minus class ZZ replacement pin gage is individually packaged, marked with size and tolerance direction, NIST traceable, and includes a Certificate of Accuracy.
  • Economical tools for quick, accurate measurements.
  • Versatile for hole sizes, distances, slot widths, and Go/NoGo gaging.
  • Designed for added wear life; features Class ZZ tolerance.
  • NIST traceable.
  • Made from tool steel (Rc 60/65) with 10 microinch or better finish.
  • Roundness within .0001 in or .0025 mm.
  • Marked with size and tolerance direction (for gages over .0605 in).
  • Includes a certificate of accuracy.
  • Overall length: 2 in.
Catalog No. 50-005-32845 Supplier MSC Supplier No. 40442626
Vermont Gage Pin Gages are economical, general-purpose tools for quick and accurate measurements of hole sizes, distances, and slot widths. They are also suitable for Go/NoGo gaging. This 15.17 mm minus class ZZ replacement pin gage is individually packaged, marked with size and tolerance direction, NIST traceable, and includes a Certificate of Accuracy.
  • Economical tools for quick, accurate measurements.
  • Versatile for hole sizes, distances, slot widths, and Go/NoGo gaging.
  • Designed for added wear life; features Class ZZ tolerance.
  • NIST traceable.
  • Made from tool steel (Rc 60/65) with 10 microinch or better finish.
  • Roundness within .0001 in or .0025 mm.
  • Marked with size and tolerance direction (for gages over .0605 in).
  • Includes a certificate of accuracy.
  • Overall length: 2 in.