Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 16.02 mm, Class ZZ, Go (Plus)
Catalog No. 5000527923
Encompass
Pin gages are essential tools for quick and accurate measurements in various applications. They can be used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. This pin gage is made from durable tool steel with a fine finish, designed for precision tasks.
  • Made from tool steel; Rc 60/65
  • Features a 10 microinch finish or better
  • Overall length of 2 in
  • Includes a certificate of accuracy with NIST traceability
  • Measures hole sizes within +.005/-.000"
  • Suitable for go/no go gaging
Catalog No. 50-005-27923 Supplier MSC Supplier No. 68107077
Pin gages are essential tools for quick and accurate measurements in various applications. They can be used to measure hole sizes, calculate distances between holes, gage slot widths, check locations, and for Go/NoGo gaging. This pin gage is made from durable tool steel with a fine finish, designed for precision tasks.
  • Made from tool steel; Rc 60/65
  • Features a 10 microinch finish or better
  • Overall length of 2 in
  • Includes a certificate of accuracy with NIST traceability
  • Measures hole sizes within +.005/-.000"
  • Suitable for go/no go gaging