Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 20.09 mm, Class ZZ, Go (Plus)
Catalog No. 5000528058
Encompass
This pin gage is engineered for the precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell C hardness of 60/65, it ensures reliable and accurate performance in various gaging applications.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Provides a tolerance of +.005/-.000.
  • Measures hole sizes with high accuracy.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length is 2 in.
  • Diameter of 20.09 mm.
  • Features a finish of 10 microfinish or better.
  • Designated as a Go (Plus) type.
  • Class ZZ rated for precision.
  • Manufactured in the United States.
Catalog No. 50-005-28058 Supplier MSC Supplier No. 68111145
This pin gage is engineered for the precise measurement of hole sizes. Constructed from durable tool steel with a Rockwell C hardness of 60/65, it ensures reliable and accurate performance in various gaging applications.
  • Constructed from tool steel with Rc 60/65 hardness.
  • Provides a tolerance of +.005/-.000.
  • Measures hole sizes with high accuracy.
  • Includes a certificate of accuracy with NIST traceability.
  • Overall length is 2 in.
  • Diameter of 20.09 mm.
  • Features a finish of 10 microfinish or better.
  • Designated as a Go (Plus) type.
  • Class ZZ rated for precision.
  • Manufactured in the United States.