Learn More
MSC Vermont Gage Plug Gage, 0.0636 in, Class X, No-Go, Steel
Catalog No. 5000533309
Encompass
This plug gage can be used as a master to calibrate measuring equipment or to gage parts directly. It provides accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Diameter of 0.0636 inches
  • No-Go (minus) direction of tolerance
  • Class X standard
  • Constructed from tool steel, cold stabilized to 60/65 Rockwell C
  • 2 inch long
  • Surface finish of 4 microinches or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
  • Tolerance of 0.00004 inches
Catalog No. 50-005-33309 Supplier MSC Supplier No. 00770131
This plug gage can be used as a master to calibrate measuring equipment or to gage parts directly. It provides accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Diameter of 0.0636 inches
  • No-Go (minus) direction of tolerance
  • Class X standard
  • Constructed from tool steel, cold stabilized to 60/65 Rockwell C
  • 2 inch long
  • Surface finish of 4 microinches or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
  • Tolerance of 0.00004 inches