Learn More
MSC Vermont Gage Plug Gage, 0.0825 in Diameter, 2.000 in OAL, Class ZZ Plus
Catalog No. 5000530558
Encompass
This plug gage is an economical and precise tool designed for quick and accurate measurements in various inspection and setting applications. It is ideal for checking hole sizes, distances between holes, and slot widths, as well as for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel with a black oxide finish.
  • Features Class ZZ tolerance for precise measurements.
  • Overall length of 2.000 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Hardness of 60 to 65 Rc for extended tool life.
  • Surface finish of 10 microinch or better.
  • Roundness within 0.0001 inches.
Catalog No. 50-005-30558 Supplier MSC Supplier No. 75872846
This plug gage is an economical and precise tool designed for quick and accurate measurements in various inspection and setting applications. It is ideal for checking hole sizes, distances between holes, and slot widths, as well as for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable tool steel with a black oxide finish.
  • Features Class ZZ tolerance for precise measurements.
  • Overall length of 2.000 inches.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Hardness of 60 to 65 Rc for extended tool life.
  • Surface finish of 10 microinch or better.
  • Roundness within 0.0001 inches.