Learn More
MSC Vermont Gage Plug Gage, 0.1015 in Diameter, 0.000200 in Tolerance, Tool Steel
Catalog No. 5000527549
Encompass
This plug gage is designed for precise measurement and inspection applications. It features a 0.1015 inch diameter and is made from durable tool steel. It is suitable for setting micrometers, verniers, and other measuring devices, and conforms to ASME B89.1 standards. Each gage includes a certificate of accuracy with NIST traceability.
  • Precise 0.1015 inch diameter
  • 0.000200 inch tolerance
  • Constructed from tool steel
  • 10 micro coating/finish
  • Ideal for go inspection applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-27549 Supplier MSC Supplier No. 88910153
This plug gage is designed for precise measurement and inspection applications. It features a 0.1015 inch diameter and is made from durable tool steel. It is suitable for setting micrometers, verniers, and other measuring devices, and conforms to ASME B89.1 standards. Each gage includes a certificate of accuracy with NIST traceability.
  • Precise 0.1015 inch diameter
  • 0.000200 inch tolerance
  • Constructed from tool steel
  • 10 micro coating/finish
  • Ideal for go inspection applications
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability