Learn More
MSC Vermont Gage Plug Gage, 0.1201 inch, Class X, Steel
Catalog No. 5000513912
Encompass
This plug gage can be used as a master to calibrate measuring equipment or to gage parts directly. It provides accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Tool steel, 60/65 Rc (cold stabilized)
  • 2 in long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
Catalog No. 50-005-13912 Supplier MSC Supplier No. 63175095
This plug gage can be used as a master to calibrate measuring equipment or to gage parts directly. It provides accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Tool steel, 60/65 Rc (cold stabilized)
  • 2 in long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included