Learn More
MSC Vermont Gage Plug Gage, 0.1253 in, Class X, Tool Steel, Go (Plus)
Catalog No. 5000528965
Encompass
These plug gages can be used as masters to calibrate measuring equipment or to gage parts directly. They provide accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Tool steel construction (60/65 Rc)
  • 2 inch long
  • 4 microinch finish or better
  • Round within half of tolerance
  • Certificate of accuracy included
  • Provides accurate, cost-effective measurement standards for shop floor and metrology laboratory use
Catalog No. 50-005-28965 Supplier MSC Supplier No. 86305711
These plug gages can be used as masters to calibrate measuring equipment or to gage parts directly. They provide accurate, cost-effective measurement standards suitable for both the shop floor and metrology laboratory.
  • Tool steel construction (60/65 Rc)
  • 2 inch long
  • 4 microinch finish or better
  • Round within half of tolerance
  • Certificate of accuracy included
  • Provides accurate, cost-effective measurement standards for shop floor and metrology laboratory use