Learn More
MSC Vermont Gage Plug Gage, 0.1955 in, Class ZZ Plus, Tool Steel, 10 Microinch Finish
Catalog No. 5000531966
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.1955 inch (4.966 mm) diameter, a 0.0002 inch (0.005 mm) tolerance, and a 10 microinch finish, making it suitable for demanding laboratory and industrial applications.
  • Diameter of 0.1955 in
  • Tolerance of 0.0002 in
  • Overall length of 2.0000 in
  • 10 microinch finish
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection
Catalog No. 50-005-31966 Supplier MSC Supplier No. 88919550
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.1955 inch (4.966 mm) diameter, a 0.0002 inch (0.005 mm) tolerance, and a 10 microinch finish, making it suitable for demanding laboratory and industrial applications.
  • Diameter of 0.1955 in
  • Tolerance of 0.0002 in
  • Overall length of 2.0000 in
  • 10 microinch finish
  • Constructed from tool steel
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection