Learn More
MSC Vermont Gage Plug Gage, 0.2080 in Diameter, 0.000200 in Tolerance, Tool Steel
Catalog No. 5000530194
Encompass
This Class ZZ Plus Plug Gage features a 0.2080 in diameter, 2.0000 in overall length, and a 0.000200 in tolerance. It has a 10 Micro finish and is constructed from tool steel. This gage is designed for go inspection, setting micrometers, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST traceability and meets ASME B89.1 standards.
  • 0.2080 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Made from tool steel
  • Features a 10 Micro finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-30194 Supplier MSC Supplier No. 88920806
This Class ZZ Plus Plug Gage features a 0.2080 in diameter, 2.0000 in overall length, and a 0.000200 in tolerance. It has a 10 Micro finish and is constructed from tool steel. This gage is designed for go inspection, setting micrometers, verniers, and other measuring devices. It includes a Certificate of Accuracy with NIST traceability and meets ASME B89.1 standards.
  • 0.2080 in diameter
  • 2.0000 in overall length
  • 0.000200 in tolerance
  • Made from tool steel
  • Features a 10 Micro finish
  • Suitable for go inspection, setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards