Learn More
MSC Vermont Gage Plug Gage, 0.23 mm, Class ZZ Plus, 2.0000 in OAL
Catalog No. 5000527958
Encompass
This Class ZZ Plus plug gage has a 0.23 mm diameter and a 2.0000 in overall length. It features a 0.00508 mm tolerance and a 10 micro finish.
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • Tool steel material
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 10 micro finish
  • 0.23 mm diameter of measurement
  • Manufactured in the United States
Catalog No. 50-005-27958 Supplier MSC Supplier No. 68091289
This Class ZZ Plus plug gage has a 0.23 mm diameter and a 2.0000 in overall length. It features a 0.00508 mm tolerance and a 10 micro finish.
  • 2.0000 in overall length
  • 0.00508 mm tolerance
  • Tool steel material
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • 10 micro finish
  • 0.23 mm diameter of measurement
  • Manufactured in the United States