Learn More
MSC Vermont Gage Plug Gage, 0.2460 in Dia, 2.0000 in OAL, Class ZZ Plus
Catalog No. 5000527544
Encompass
This Class ZZ Plus plug gage, constructed from durable tool steel, features a diameter of 0.2460 in and an overall length of 2.0000 in. It is suitable for go inspection and setting micrometers, verniers, and other measuring devices, providing precise measurements with a 0.000200 in tolerance.
  • Certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-27544 Supplier MSC Supplier No. 88924600
This Class ZZ Plus plug gage, constructed from durable tool steel, features a diameter of 0.2460 in and an overall length of 2.0000 in. It is suitable for go inspection and setting micrometers, verniers, and other measuring devices, providing precise measurements with a 0.000200 in tolerance.
  • Certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 micro coating/finish.