Learn More
MSC Vermont Gage Plug Gage, 0.2610 in Dia, 2.0000 in OAL
Catalog No. 5000530225
Encompass
This precision plug gage is designed for demanding inspection and calibration tasks. It is ideal for Go inspection and setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this gage provides accuracy and reliability for a wide range of applications.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Made from high-quality tool steel
  • Features a 10 micro coating/finish
  • Proudly made in the United States
Catalog No. 50-005-30225 Supplier MSC Supplier No. 88926100
This precision plug gage is designed for demanding inspection and calibration tasks. It is ideal for Go inspection and setting micrometers, verniers, and other measuring devices. Constructed from durable tool steel, this gage provides accuracy and reliability for a wide range of applications.
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
  • Made from high-quality tool steel
  • Features a 10 micro coating/finish
  • Proudly made in the United States