Learn More
MSC Vermont Gage Plug Gage, 0.338", Class ZZ Plus, Go, Tool Steel
Catalog No. 5000515148
Encompass
This plug gage is designed for precise 'Go' inspection applications. It features a 0.338 inch diameter with a Class ZZ Plus tolerance of 0.0002 inch and a 10 microfinish surface. It is ideal for verifying hole sizes and ensuring manufacturing precision.
  • Designed for go inspection
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from durable tool steel
  • Conforms to ASME B89.1 standards
  • Features a 10 microfinish
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inch
  • Measurement diameter of 0.3380 inch
Catalog No. 50-005-15148 Supplier MSC Supplier No. 88933809
This plug gage is designed for precise 'Go' inspection applications. It features a 0.338 inch diameter with a Class ZZ Plus tolerance of 0.0002 inch and a 10 microfinish surface. It is ideal for verifying hole sizes and ensuring manufacturing precision.
  • Designed for go inspection
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from durable tool steel
  • Conforms to ASME B89.1 standards
  • Features a 10 microfinish
  • Overall length of 2.0000 inches
  • Tolerance of 0.000200 inch
  • Measurement diameter of 0.3380 inch