Learn More
MSC Vermont Gage Plug Gage, 0.3500 in x 2.0000 in, Tool Steel
Catalog No. 5000530534
Encompass
This plug gage is designed for precise measurement and inspection tasks, featuring a 0.3500 in diameter and 2.0000 in overall length. Constructed from durable tool steel, it is ideal for checking dimensions, calibrating instruments, and ensuring quality control in various applications.
  • Certificate of accuracy with NIST traceability included
  • Adheres to ASME B89.1 standards
  • Coated with a 10 micro finish
  • Ideal for go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Made from robust tool steel
Catalog No. 50-005-30534 Supplier MSC Supplier No. 88935002
This plug gage is designed for precise measurement and inspection tasks, featuring a 0.3500 in diameter and 2.0000 in overall length. Constructed from durable tool steel, it is ideal for checking dimensions, calibrating instruments, and ensuring quality control in various applications.
  • Certificate of accuracy with NIST traceability included
  • Adheres to ASME B89.1 standards
  • Coated with a 10 micro finish
  • Ideal for go inspection
  • Can be used to set micrometers, verniers, and other measuring devices
  • Made from robust tool steel