Learn More
MSC Vermont Gage Plug Gage, 0.3665 in Diameter, 0.0002 in Tolerance, 10 Microfinish
Catalog No. 5000515615
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.3665 in diameter with a tight 0.0002 in tolerance and a 10 microfinish. Made from durable tool steel, it is suitable for various industrial applications.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 microfinish for precision.
  • Made from tool steel for durability.
Catalog No. 50-005-15615 Supplier MSC Supplier No. 88936653
This Class ZZ Plus plug gage is designed for precise measurement and inspection. It features a 0.3665 in diameter with a tight 0.0002 in tolerance and a 10 microfinish. Made from durable tool steel, it is suitable for various industrial applications.
  • For go inspection, setting micrometers, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured to ASME B89.1 standards.
  • Features a 10 microfinish for precision.
  • Made from tool steel for durability.