Learn More
MSC Vermont Gage Plug Gage, 0.3898 in, Class X, Go (Plus)
Catalog No. 5000529733
Encompass
This standard Class X plug gage is designed for precise measurement applications, serving as a master to calibrate measuring equipment or to directly gage parts. It offers accurate, cost-effective measurement standards suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance: +0.00004 in, -0.0000 in
  • NIST traceable
  • Tool steel construction; 60/65 Rc (cold stabilized)
  • 2 in long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
  • Go (Plus) tolerance type
Catalog No. 50-005-29733 Supplier MSC Supplier No. 06313530
This standard Class X plug gage is designed for precise measurement applications, serving as a master to calibrate measuring equipment or to directly gage parts. It offers accurate, cost-effective measurement standards suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance: +0.00004 in, -0.0000 in
  • NIST traceable
  • Tool steel construction; 60/65 Rc (cold stabilized)
  • 2 in long
  • 4 microinch finish or better
  • Round within 1/2 of tolerance
  • Certificate of accuracy included
  • Go (Plus) tolerance type