Learn More
MSC Vermont Gage Plug Gage, 0.4005 inch, Class ZZ Plus, Tool Steel
Catalog No. 5000536737
Encompass
A Class ZZ Plus Plug Gage with a diameter of 0.4005 inches and an overall length of 2.0000 inches, featuring a tolerance of 0.000200 inches. It is made of tool steel and is intended for "Go" inspection and setting various measuring devices. This gage meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
  • Made of tool steel.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-36737 Supplier MSC Supplier No. 88940051
A Class ZZ Plus Plug Gage with a diameter of 0.4005 inches and an overall length of 2.0000 inches, featuring a tolerance of 0.000200 inches. It is made of tool steel and is intended for "Go" inspection and setting various measuring devices. This gage meets ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability.
  • Made of tool steel.
  • Suitable for go inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Meets ASME B89.1 standards.
  • Features a 10 micro coating/finish.