Learn More
MSC Vermont Gage Plug Gage, 0.4210 in Diameter, 2.0000 in Length, Tool Steel, ASME B89.1
Catalog No. 5000531722
Encompass
This plug gage is designed for precise measurement and inspection, featuring a 0.4210 in diameter and a 2.0000 in overall length. Constructed from durable tool steel with a 10 micro finish, it is ideal for go inspection and setting various measuring devices. It conforms to ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability
  • Hassle free guarantee
Catalog No. 50-005-31722 Supplier MSC Supplier No. 88942107
This plug gage is designed for precise measurement and inspection, featuring a 0.4210 in diameter and a 2.0000 in overall length. Constructed from durable tool steel with a 10 micro finish, it is ideal for go inspection and setting various measuring devices. It conforms to ASME B89.1 standards.
  • Includes certificate of accuracy with NIST traceability
  • Hassle free guarantee