Learn More
MSC Vermont Gage Plug Gage, 0.4270 in Diameter, 2.0000 in OAL, Tool Steel
Catalog No. 5000531970
Encompass
This Class ZZ Plus Plug Gage is crafted from tool steel and offers high precision for various measurement applications. With a diameter of 0.4270 inch and an overall length of 2.0000 inches, it features a tolerance of 0.000200 inch, ensuring accurate gauging.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-31970 Supplier MSC Supplier No. 88942701
This Class ZZ Plus Plug Gage is crafted from tool steel and offers high precision for various measurement applications. With a diameter of 0.4270 inch and an overall length of 2.0000 inches, it features a tolerance of 0.000200 inch, ensuring accurate gauging.
  • For go inspection, setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability