Learn More
MSC Vermont Gage Plug Gage, 0.4395 in, 0.0002 in Tolerance, 10 Microfinish
Catalog No. 5000515395
Encompass
This plug gage is designed for go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.4395 inch diameter, 0.0002 inch tolerance, and a 10 microfinish. Constructed from tool steel, this gage ensures reliable measurements and conforms to industry standards.
  • Certificate of accuracy with NIST traceability included.
  • Manufactured to ASME B89.1 standards.
  • Tool steel material for durability.
Catalog No. 50-005-15395 Supplier MSC Supplier No. 88943956
This plug gage is designed for go inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.4395 inch diameter, 0.0002 inch tolerance, and a 10 microfinish. Constructed from tool steel, this gage ensures reliable measurements and conforms to industry standards.
  • Certificate of accuracy with NIST traceability included.
  • Manufactured to ASME B89.1 standards.
  • Tool steel material for durability.