Learn More
MSC Vermont Gage Plug Gage, 0.5010 in Diameter, 2.0000 in OAL, Class ZZ Plus
Catalog No. 5000531724
Encompass
This Vermont Gage Class ZZ Plus Plug Gage is engineered for high-precision measurement and calibration tasks. With a precise 0.5010 in diameter and a 2.0000 in overall length, this tool steel gage offers reliable performance for setting micrometers, verniers, and other measuring devices, as well as for go inspection.
  • Features a class ZZ Plus tolerance.
  • Equipped with a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.
Catalog No. 50-005-31724 Supplier MSC Supplier No. 88950100
This Vermont Gage Class ZZ Plus Plug Gage is engineered for high-precision measurement and calibration tasks. With a precise 0.5010 in diameter and a 2.0000 in overall length, this tool steel gage offers reliable performance for setting micrometers, verniers, and other measuring devices, as well as for go inspection.
  • Features a class ZZ Plus tolerance.
  • Equipped with a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Complies with ASME B89.1 standards.