Learn More
MSC Vermont Gage Plug Gage, 0.5090 in, 0.000200 in Tolerance
Catalog No. 5000515073
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurements and calibration in various applications. It features a 0.5090 in diameter with a 0.000200 in tolerance and a 10 Micro finish. Constructed from durable tool steel, it is suitable for go inspection and for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Class ZZ Plus plug gage for precise measurement.
  • 0.5090 in diameter.
  • 2.0000 in overall length.
  • 0.000200 in tolerance.
  • Constructed from durable tool steel.
  • Features a 10 Micro finish.
  • Suitable for go inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Manufactured in the United States.
Catalog No. 50-005-15073 Supplier MSC Supplier No. 88950902
This Class ZZ Plus Plug Gage is designed for precise measurements and calibration in various applications. It features a 0.5090 in diameter with a 0.000200 in tolerance and a 10 Micro finish. Constructed from durable tool steel, it is suitable for go inspection and for setting micrometers, verniers, and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability and conforms to ASME B89.1 standards.
  • Class ZZ Plus plug gage for precise measurement.
  • 0.5090 in diameter.
  • 2.0000 in overall length.
  • 0.000200 in tolerance.
  • Constructed from durable tool steel.
  • Features a 10 Micro finish.
  • Suitable for go inspection and setting various measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Conforms to ASME B89.1 standards.
  • Manufactured in the United States.