Learn More
MSC Vermont Gage Plug Gage, 0.6490 in, 0.0002 in Tolerance, Tool Steel
Catalog No. 5000536555
Encompass
This Class ZZ Plus Plug Gage is crafted from durable tool steel, designed for precise measurements and inspection. With a diameter of 0.6490 inch and an overall length of 2.0000 inch, it features a tolerance of 0.0002 inch and a 10 microinch finish, ensuring accuracy in various applications. It includes a certificate of accuracy with NIST traceability and meets ASME B89.1 standards.
  • For quick and accurate measurements
  • Constructed from tool steel
  • 0.6490 inch diameter
  • 2.0000 inch overall length
  • Tolerance of 0.0002 inch
  • 10 microinch finish
  • Suitable for Go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-36555 Supplier MSC Supplier No. 88964903
This Class ZZ Plus Plug Gage is crafted from durable tool steel, designed for precise measurements and inspection. With a diameter of 0.6490 inch and an overall length of 2.0000 inch, it features a tolerance of 0.0002 inch and a 10 microinch finish, ensuring accuracy in various applications. It includes a certificate of accuracy with NIST traceability and meets ASME B89.1 standards.
  • For quick and accurate measurements
  • Constructed from tool steel
  • 0.6490 inch diameter
  • 2.0000 inch overall length
  • Tolerance of 0.0002 inch
  • 10 microinch finish
  • Suitable for Go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards