Learn More
MSC Vermont Gage Plug Gage, 0.74 mm, Class ZZ Plus, Tool Steel
Catalog No. 5000531049
Encompass
This Class ZZ Plus Plug Gage is designed for precision measurement, featuring a 0.74 mm diameter and constructed from durable tool steel for accuracy and longevity. It is suitable for various inspection tasks, including 'Go' inspection and setting other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • For go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Tolerance of 0.00508 mm.
  • 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Overall length of 2.0000 in.
Catalog No. 50-005-31049 Supplier MSC Supplier No. 68091792
This Class ZZ Plus Plug Gage is designed for precision measurement, featuring a 0.74 mm diameter and constructed from durable tool steel for accuracy and longevity. It is suitable for various inspection tasks, including 'Go' inspection and setting other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • For go inspection.
  • Sets micrometers, verniers, and other measuring devices.
  • Tolerance of 0.00508 mm.
  • 10 micro coating/finish.
  • Conforms to ASME B89.1 standards.
  • Overall length of 2.0000 in.