Learn More
MSC Vermont Gage Plug Gage, 0.8250 in Diameter, 2.0000 in Length, Tool Steel
Catalog No. 5000513693
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications, including "Go" inspection and setting measuring devices such as micrometers and verniers. Constructed from durable tool steel, it features a 0.8250 in diameter and an overall length of 2.0000 in, ensuring accuracy with a tolerance of 0.000200 in.
  • Designed for "Go" inspection and setting measuring devices.
  • Constructed from durable tool steel.
  • Features a 0.8250 in diameter.
  • Measures 2.0000 in in overall length.
  • Maintains an accurate tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Features a 10 micro coating/finish.
Catalog No. 50-005-13693 Supplier MSC Supplier No. 88982509
This Class ZZ Plus Plug Gage is designed for precise measurement applications, including "Go" inspection and setting measuring devices such as micrometers and verniers. Constructed from durable tool steel, it features a 0.8250 in diameter and an overall length of 2.0000 in, ensuring accuracy with a tolerance of 0.000200 in.
  • Designed for "Go" inspection and setting measuring devices.
  • Constructed from durable tool steel.
  • Features a 0.8250 in diameter.
  • Measures 2.0000 in in overall length.
  • Maintains an accurate tolerance of 0.000200 in.
  • Includes a certificate of accuracy with NIST traceability.
  • Adheres to ASME B89.1 standards.
  • Features a 10 micro coating/finish.