Learn More
MSC Vermont Gage Plug Gage, 0.86 mm, Class ZZ Plus, Black Oxide
Catalog No. 5000527292
Encompass
This plug gage is designed for precise measurement applications. It features a 0.86 mm diameter and a black oxide finish, providing durability and accuracy for various industrial uses. The gage ensures consistent performance with its Class ZZ Plus tolerance.
  • 0.86 mm diameter
  • 2.0000 in overall length
  • Class ZZ Plus tolerance of 0.00508 mm
  • Black oxide finish
  • Manufactured in the USA
Catalog No. 50-005-27292 Supplier MSC Supplier No. 65922544
This plug gage is designed for precise measurement applications. It features a 0.86 mm diameter and a black oxide finish, providing durability and accuracy for various industrial uses. The gage ensures consistent performance with its Class ZZ Plus tolerance.
  • 0.86 mm diameter
  • 2.0000 in overall length
  • Class ZZ Plus tolerance of 0.00508 mm
  • Black oxide finish
  • Manufactured in the USA