Learn More
MSC Vermont Gage Plug Gage, 0.890 inch, Class ZZ Minus, Tool Steel
Catalog No. 5000531664
Encompass
This plug gage is designed for precise measurement and verification, featuring a 0.890 inch diameter. Crafted from durable tool steel, it ensures reliability and accuracy in gaging applications.
  • Class ZZ tolerance provides high accuracy for precise gaging.
  • Made from durable tool steel with 60/65 Rc hardness.
  • 2-inch long for comfortable handling.
  • Features a 10 microinch finish or better for smooth operation.
  • Round within 0.0001 inch for consistent measurements.
  • Marked with diameter and direction of tolerance on gages over 0.0605 "
  • Includes a certificate of accuracy.
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased.
Catalog No. 50-005-31664 Supplier MSC Supplier No. 78380862
This plug gage is designed for precise measurement and verification, featuring a 0.890 inch diameter. Crafted from durable tool steel, it ensures reliability and accuracy in gaging applications.
  • Class ZZ tolerance provides high accuracy for precise gaging.
  • Made from durable tool steel with 60/65 Rc hardness.
  • 2-inch long for comfortable handling.
  • Features a 10 microinch finish or better for smooth operation.
  • Round within 0.0001 inch for consistent measurements.
  • Marked with diameter and direction of tolerance on gages over 0.0605 "
  • Includes a certificate of accuracy.
  • ISO 17025 accredited and non-accredited calibration certificates may be purchased.