Learn More
MSC Vermont Gage Plug Gage, 0.9170 in Diameter, Tool Steel, 0.000200 in Tolerance
Catalog No. 5000531571
Encompass
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks. Constructed from durable tool steel, it features a 0.9170 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 "
  • Designed for Go inspection and setting various measuring devices
  • Constructed from tool steel for enhanced durability
  • Features a 10 micro coating/finish for consistent performance
  • Complies with ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability for assured quality
  • Manufactured in the United States
Catalog No. 50-005-31571 Supplier MSC Supplier No. 88991708
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks. Constructed from durable tool steel, it features a 0.9170 inch diameter and a 2.0000 inch overall length, with a tight tolerance of 0.000200 "
  • Designed for Go inspection and setting various measuring devices
  • Constructed from tool steel for enhanced durability
  • Features a 10 micro coating/finish for consistent performance
  • Complies with ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability for assured quality
  • Manufactured in the United States