Learn More
MSC Vermont Gage Plug Gage, 1.2889 in, Class ZZ Plus, Go (Plus)
Catalog No. 5000513809
Encompass
This Class ZZ Plus Plug Gage is an economical, general-purpose pin gage designed for quick and accurate measurements. With a diameter of 1.2889 in and a Class ZZ tolerance of .0002 in, it is suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging. This 'Go (Plus)' gage provides added wear life.
  • Handle sold separately.
  • NIST traceable.
  • Certificate of accuracy included.
  • Calibration services are available.
  • Length: 2 in.
  • Finish of 10 microinch or better.
  • Round within .0001 in.
  • Marked with size and direction of tolerance on gages over .0605 in.
Catalog No. 50-005-13809 Supplier MSC Supplier No. 64758006
This Class ZZ Plus Plug Gage is an economical, general-purpose pin gage designed for quick and accurate measurements. With a diameter of 1.2889 in and a Class ZZ tolerance of .0002 in, it is suitable for measuring hole sizes, calculating distances between holes, gaging slot widths, checking locations, and Go/NoGo gaging. This 'Go (Plus)' gage provides added wear life.
  • Handle sold separately.
  • NIST traceable.
  • Certificate of accuracy included.
  • Calibration services are available.
  • Length: 2 in.
  • Finish of 10 microinch or better.
  • Round within .0001 in.
  • Marked with size and direction of tolerance on gages over .0605 in.