Learn More
MSC Vermont Gage Plug Gage, 1.2931 in Diameter, Class ZZ Plus
Catalog No. 5000530960
Encompass
This plug gage is a precision measuring tool designed for accurate verification of hole sizes. Engineered with a 1.2931 inch diameter and Class ZZ Plus tolerance, it provides reliable results for quality inspection and manufacturing processes.
  • Precision plug gage for internal diameter measurement.
  • 1.2931 in diameter.
  • Features Class ZZ Plus tolerance.
  • Manufactured in the United States.
Catalog No. 50-005-30960 Supplier MSC Supplier No. 64758428
This plug gage is a precision measuring tool designed for accurate verification of hole sizes. Engineered with a 1.2931 inch diameter and Class ZZ Plus tolerance, it provides reliable results for quality inspection and manufacturing processes.
  • Precision plug gage for internal diameter measurement.
  • 1.2931 in diameter.
  • Features Class ZZ Plus tolerance.
  • Manufactured in the United States.