Learn More
MSC Vermont Gage Plug Gage, 1.2967 in, Class ZZ Plus, Tool Steel
Catalog No. 5000530322
Encompass
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate hole inspection. It features a 1.2967 inch diameter and a 0.0002 inch tolerance, ensuring high accuracy for quality control applications.
  • Made from durable tool steel
  • Features a black oxide finish
  • Provides a 10 microinch surface finish
  • Handle sold separately for customizable use
Catalog No. 50-005-30322 Supplier MSC Supplier No. 64758782
The Vermont Gage Class ZZ Plus Plug Gage is a precision measuring tool designed for accurate hole inspection. It features a 1.2967 inch diameter and a 0.0002 inch tolerance, ensuring high accuracy for quality control applications.
  • Made from durable tool steel
  • Features a black oxide finish
  • Provides a 10 microinch surface finish
  • Handle sold separately for customizable use