Learn More
MSC Vermont Gage Plug Gage, 1.84 mm Diameter, 2.00 in OAL, 10 Micro Finish
Catalog No. 5000529019
Encompass
This Class ZZ Plus Plug Gage is engineered for precision measurement tasks, featuring a 1.84 mm diameter, a tight tolerance of 0.00508 mm, and a smooth 10 Micro finish. It is specifically designed for 'Go Inspection' applications and for accurately setting various measuring instruments like micrometers and verniers. Each gage comes with a Certificate of Accuracy, ensuring NIST traceability, and complies with ASME B89.1 standards.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Made from durable tool steel
  • Features a 10 micro finish
  • Has an overall length of 2.00 inches
Catalog No. 50-005-29019 Supplier MSC Supplier No. 68092899
This Class ZZ Plus Plug Gage is engineered for precision measurement tasks, featuring a 1.84 mm diameter, a tight tolerance of 0.00508 mm, and a smooth 10 Micro finish. It is specifically designed for 'Go Inspection' applications and for accurately setting various measuring instruments like micrometers and verniers. Each gage comes with a Certificate of Accuracy, ensuring NIST traceability, and complies with ASME B89.1 standards.
  • Designed for go inspection
  • Used to set micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
  • Made from durable tool steel
  • Features a 10 micro finish
  • Has an overall length of 2.00 inches