Learn More
MSC Vermont Gage Plug Gage, 1.86 mm, Class ZZ Plus, Tool Steel, 10 Micro
Catalog No. 5000530965
Encompass
This Vermont Gage plug gage is designed for precise measurement and inspection applications. It features a 1.86 mm diameter and is constructed from durable tool steel with a 10 micro finish, ensuring accuracy and longevity.
  • Includes certificate of accuracy with NIST traceability.
  • For go inspection.
  • Set micrometers, verniers, and other measuring devices.
Catalog No. 50-005-30965 Supplier MSC Supplier No. 68092915
This Vermont Gage plug gage is designed for precise measurement and inspection applications. It features a 1.86 mm diameter and is constructed from durable tool steel with a 10 micro finish, ensuring accuracy and longevity.
  • Includes certificate of accuracy with NIST traceability.
  • For go inspection.
  • Set micrometers, verniers, and other measuring devices.